Merck
  • IRMS detection of testosterone manipulated with 13C labeled standards in human urine by removing the labeled 13C.
IRMS detection of testosterone manipulated with 13C labeled standards in human urine by removing the labeled 13C.
Analytica chimica acta (2014-12-03)
Jingzhu Wang, Rui Yang, Wenning Yang, Xin Liu, Yanyi Xing, Youxuan Xu
ABSTRACT
Isotope ratio mass spectrometry (IRMS) is applied to confirm testosterone (T) abuse by determining the carbon isotope ratios (δ(13)C value). However, (13)C labeled standards can be used to control the δ(13)C value and produce manipulated T which cannot be detected by the current method. A method was explored to remove the (13)C labeled atom at C-3 from the molecule of androsterone (Andro), the metabolite of T in urine, to produce the resultant (A-nor-5α-androstane-2,17-dione, ANAD). The difference in δ(13)C values between Andro and ANAD (Δδ(13)CAndro-ANAD, ‰) would change significantly in case manipulated T is abused. Twenty-one volunteers administered T manipulated with different (13)C labeled standards. The collected urine samples were analyzed with the established method, and the maximum value of Δδ(13)CAndro-ANAD post ingestion ranged from 3.0‰ to 8.8‰. Based on the population reference, the cut-off value of Δδ(13)CAndro-ANAD for positive result was suggested as 1.2‰. The developed method could be used to detect T manipulated with 3-(13)C labeled standards.
Product Description
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Sigma-Aldrich
17α-Methyltestosterone, solid (photosensitive)
Methyltestosterone, European Pharmacopoeia (EP) Reference Standard
Methyltestosterone for system suitability, European Pharmacopoeia (EP) Reference Standard