Merck
  • Characterisation of triterpenes and new phenolic lipids in Cameroonian propolis.
Characterisation of triterpenes and new phenolic lipids in Cameroonian propolis.
Phytochemistry (2014-08-12)
M N Kardar, T Zhang, G D Coxon, D G Watson, J Fearnley, V Seidel
ABSTRACT
Chemical investigation of a sample of propolis originating from North-Western Cameroon led to the isolation of thirteen alk(en)ylphenols (1-13) (inseparable mixture) along with α-amyrin (14), β-amyrin (15), lupeol (16), cycloartenol (17), mangiferonic acid (18), ambonic acid (19), mangiferolic acid (20), ambolic acid (21), isomangiferolic acid (22) and nine alk(en)ylresorcinols (23-31) (inseparable mixture). All compounds were identified following analysis of their spectroscopic data and comparison with previously published reports. Compounds (8), (12), (13) and (30) are new natural products. GC-MS analysis carried out on the alk(en)ylphenol and alk(en)ylresorcinol mixtures (dimethyl disulphide trimethylsilyl derivatives) revealed the presence of saturated and mono-unsaturated compounds with side chain lengths ranging from C11 to C19 and C15 to C19, respectively. The position of the double bond in mono-unsaturated derivatives was established from the characteristic fragments resulting from the cleavage of the bond between the two methylthio-substituted carbons. The most abundant compound in each mixture was 3-(12'Z-heptadecenyl)-phenol (10) and 5-(12'Z-heptadecenyl)-resorcinol (29). This study is the first to report the presence of triterpenes (except for lupeol) and phenolic lipids, including eighteen compounds previously unreported in bee glue, in an African sample.
Sigma-Aldrich
Resorcinol, JIS special grade, ≥99.0%
Supelco
5-Heptadecylresorcinol, analytical standard
Sigma-Aldrich
Cycloartenol, ≥90% (GC)
Supelco
Resorcinol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
USP
Resorcinol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Resorcinol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Resorcinol, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Hymecromone impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
p-Anisaldehyde, 98%
Sigma-Aldrich
Dimethyl disulfide, ≥98%, FG
Sigma-Aldrich
Trifluoroacetamide, 97%
Sigma-Aldrich
Orcinol, 97%
Sigma-Aldrich
Dimethyl disulfide, ≥99.0%
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Supelco
p-Anisaldehyde, analytical standard
Supelco
5-Pentadecylresorcinol, analytical standard
Supelco
Dimethyl disulfide, analytical standard
Sigma-Aldrich
p-Anisaldehyde, natural, FG