Merck
  • Differential activity of stanniocalcin in male and female fresh water teleost Mastacembelus armatus (Lacepede) during gonadal maturation.
Differential activity of stanniocalcin in male and female fresh water teleost Mastacembelus armatus (Lacepede) during gonadal maturation.
PloS one (2014-07-09)
Sushant Kumar Verma, Abdul Alim
ABSTRACT
The present study was carried out to analyze the differences in the activity of hormone stanniocalcin (STC) between male and female fishes of Mastacembelus armatus during their gonadal cycle. A large variation in nuclear diameter of cells of corpuscles of Stannius (CS) were recorded in relation to testicular cycle as well as ovarian cycle which indicates that the cellular activity varied with different phases of reproductive cycle in both male and female fish. Similar changes in nuclear diameter of CS cells were also observed after 17alpha-methyltestosterone administration in males and 17 β-estradiol administrations in females. A positive correlation was observed between plasma STC levels, gonadosomatic index (GSI) and the sex steroids in both sexes, suggesting that STC has a role in the processes involved in gonadal development. In addition females showed remarkable changes in plasma calcium level during gonadal cycle while no such change for males were observed. In females the plasma calcium level estimated during different phases of reproductive cycle indicates positive correlation between plasma level of calcium and gonad growth. Thus hyperactivity of CS cells was noted in both male and female fishes during gonadal cycle along with the differences in the activity of STC as well. In female it may act as hypocalcemic factor and bring the level of calcium to normal which increases during preparatory and pre spawning phases to fulfill the increased demand of calcium for vitellogenesis. However data of male fishes indicated that plasma STC concentration varied widely during gonadal cycle but showed no consistent relationship to plasma calcium level.
Sigma-Aldrich
17α-Methyltestosterone, solid (photosensitive)
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Methyltestosterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Methyltestosterone, European Pharmacopoeia (EP) Reference Standard