Merck
  • Synthesis of tetra-substituted imidazoles and 2-imidazolines by Ni(0)-catalyzed dehydrogenation of benzylic-type imines.
Synthesis of tetra-substituted imidazoles and 2-imidazolines by Ni(0)-catalyzed dehydrogenation of benzylic-type imines.
Dalton transactions (Cambridge, England : 2003) (2014-09-19)
Adrian Tlahuext-Aca, Oscar Hernández-Fajardo, Alma Arévalo, Juventino J García
ABSTRACT
Ni(0)-catalyzed dehydrogenation of benzylic-type imines was performed to yield asymmetrical tetra-substituted imidazoles and 2-imidazolines. This was achieved with a single operational step while maintaining good selectivity and atom economy. The catalytic system shows low to moderate tolerance for fluoro-, trifluoromethyl-, methyl-, and methoxy-substituted benzylic-type imines. In addition, the substitution pattern at the N-heterocyclic products was easily controlled by the appropriate selection of R-groups in the starting organic substrates. Based on experimental observations, we propose a reaction mechanism in which benzylic C(sp(3))-H bond activation and insertion steps play pivotal roles in this nickel-catalyzed organic transformation.
Sigma-Aldrich
Ethane, 99.99%
Sigma-Aldrich
Tricyclohexylphosphine solution, 20 wt. % in toluene
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Ethane, ≥99%
Sigma-Aldrich
Triphenyl phosphite, 97%
Sigma-Aldrich
1,5-Cyclooctadiene, contains 50-150 ppm 4-tert-Butylcatechol, 99%
Sigma-Aldrich
1,5-Cyclooctadiene, purified by redistillation, ≥99%
Sigma-Aldrich
Bis(1,5-cyclooctadiene)nickel(0)
Sigma-Aldrich
Tricyclohexylphosphine
Sigma-Aldrich
Argon-40Ar, 99.95 atom %