Merck
Hepatic-targeted gene delivery using cationic mannan vehicle.
Molecular pharmaceutics (2014-04-17)
Gui-Xin Ruan, Tian-Yuan Zhang, Li-Ming Li, Xing-Guo Zhang, You-Qing Shen, Yasuhiko Tabata, Jian-Qing Gao
ABSTRACT
The incidence of hepatic diseases continuously increases worldwide and causes significant mortality because of inefficient pharmacotherapy. Gene therapy is a new strategy in the treatment of hepatic diseases, but the disadvantages of insufficient retention in the liver and undesirable side effects hinder its application. In this study, we developed a novel nonviral vehicle targeted to liver. Mannan was cationized with spermine at varying grafted ratios to deliver the gene and was optimized in cytotoxicity and transfection in vitro. A spermine-mannan (SM)-based delivery system was proven to be hepatic targeted and was capable of prolonging the gene retention period in the liver. Moreover, SM at N/P of 20 was confirmed to be less interfered with by the serum. Optimized SM vehicle has relatively high hepatic transfection with almost no toxicity induction in the liver, which highlighted its potential in the treatment of hepatic diseases.
Sigma-Aldrich
Spermine, ≥99.0% (GC)
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
Spermine, suitable for cell culture, BioReagent
Supelco
Spermine, analytical standard