Merck
SIRT6 is required for normal retinal function.
PloS one (2014-06-05)
Dafne M Silberman, Kenneth Ross, Pablo H Sande, Shunsuke Kubota, Sridhar Ramaswamy, Rajendra S Apte, Raul Mostoslavsky
ABSTRACT
The retina is one of the major energy consuming tissues within the body. In this context, synaptic transmission between light-excited rod and cone photoreceptors and downstream ON-bipolar neurons is a highly demanding energy consuming process. Sirtuin 6 (SIRT6), a NAD-dependent deacylase, plays a key role in regulating glucose metabolism. In this study, we demonstrate that SIRT6 is highly expressed in the retina, controlling levels of histone H3K9 and H3K56 acetylation. Notably, despite apparent normal histology, SIRT6 deficiency caused major retinal transmission defects concomitant to changes in expression of glycolytic genes and glutamate receptors, as well as elevated levels of apoptosis in inner retina cells. Our results identify SIRT6 as a critical modulator of retinal function, likely through its effects on chromatin.
Sigma-Aldrich
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
Supelco
Atropine Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Atropine sulfate, United States Pharmacopeia (USP) Reference Standard
Atropine sulfate, European Pharmacopoeia (EP) Reference Standard