Merck
  • Lewis acidity enhancement of triarylborane by appended phosphine oxide groups.
Lewis acidity enhancement of triarylborane by appended phosphine oxide groups.
Dalton transactions (Cambridge, England : 2003) (2015-02-11)
Jaewoo Kwak, Nguyen Van Nghia, Junseong Lee, Hyoseok Kim, Myung Hwan Park, Min Hyung Lee
ABSTRACT
A series of mono-, di-, and tri-phosphine oxide substituted triarylboranes, Mes2BAr (1), MesBAr2 (2), and BAr3 (3) (Ar = 4-(Ph2PO)-2,6-Me2-C6H2) were prepared to investigate the effect of a phosphine oxide group (Ph2PO) on Lewis acidity enhancement of triarylboranes. The X-ray crystal structure of 3 revealed peripheral decoration of Ph2PO groups with a C3-axis perpendicular to the trigonal boron center. UV/Vis absorption and PL spectra indicated a significant contribution of π(Mes or phenylene) → pπ(B) charge transfer in the lower-energy electronic transition. The reduction potential measured by cyclic voltammetry showed apparent LUMO stabilization by introduction of phosphine oxide groups, the extent of which gradually increased with the increasing number of phosphine oxide groups. Lewis acidity enhancement was also supported by the gradual increase in fluoride ion affinity in the order 3 > 2 > 1. Theoretical calculations suggest that introduction of a Ph2PO group into a triarylborane significantly enhances the Lewis acidity of the boron center via an inductive electron-withdrawing effect and this effect is additive for multiple phosphine oxide groups.
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Chlorodiphenylphosphine, 96%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
P-Chlorodiphenylphosphine, 99%