Merck
  • Interactions of endosulfan and methoxychlor involving CYP3A4 and CYP2B6 in human HepaRG cells.
Interactions of endosulfan and methoxychlor involving CYP3A4 and CYP2B6 in human HepaRG cells.
Drug metabolism and disposition: the biological fate of chemicals (2014-05-17)
Camille C Savary, Rozenn Jossé, Arnaud Bruyère, Fabrice Guillet, Marie-Anne Robin, André Guillouzo
ABSTRACT
Humans are usually exposed to several pesticides simultaneously; consequently, combined actions between pesticides themselves or between pesticides and other chemicals need to be addressed in the risk assessment. Many pesticides are efficient activators of pregnane X receptor (PXR) and/or constitutive androstane receptor (CAR), two major nuclear receptors that are also activated by other substrates. In the present work, we searched for interactions between endosulfan and methoxychlor, two organochlorine pesticides whose major routes of metabolism involve CAR- and PXR-regulated CYP3A4 and CYP2B6, and whose mechanisms of action in humans remain poorly understood. For this purpose, HepaRG cells were treated with both pesticides separately or in mixture for 24 hours or 2 weeks at concentrations relevant to human exposure levels. In combination they exerted synergistic cytotoxic effects. Whatever the duration of treatment, both compounds increased CYP3A4 and CYP2B6 mRNA levels while differently affecting their corresponding activities. Endosulfan exerted a direct reversible inhibition of CYP3A4 activity that was confirmed in human liver microsomes. By contrast, methoxychlor induced this activity. The effects of the mixture on CYP3A4 activity were equal to the sum of those of each individual compound, suggesting an additive effect of each pesticide. Despite CYP2B6 activity being unchanged and increased with endosulfan and methoxychlor, respectively, no change was observed with their mixture, supporting an antagonistic effect. Altogether, our data suggest that CAR and PXR activators endosulfan and methoxychlor can interact together and with other exogenous substrates in human hepatocytes. Their effects on CYP3A4 and CYP2B6 activities could have important consequences if extrapolated to the in vivo situation.
Product Description
Nifedipine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Oxidized Nifedipine, powder, ~95% (HPLC)
USP
Nifedipine Nitrophenylpyridine Analog, United States Pharmacopeia (USP) Reference Standard
USP
Nifedipine, United States Pharmacopeia (USP) Reference Standard
Supelco
Nifedipine, Pharmaceutical Secondary Standard; Certified Reference Material
Nifedipine impurity A, European Pharmacopoeia (EP) Reference Standard
Supelco
Endosulfan, PESTANAL®, analytical standard
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material