Merck
  • Neoadjuvant antiangiogenic therapy reveals contrasts in primary and metastatic tumor efficacy.
Neoadjuvant antiangiogenic therapy reveals contrasts in primary and metastatic tumor efficacy.
EMBO molecular medicine (2014-11-02)
John M L Ebos, Michalis Mastri, Christina R Lee, Amanda Tracz, John M Hudson, Kristopher Attwood, William R Cruz-Munoz, Christopher Jedeszko, Peter Burns, Robert S Kerbel
ABSTRACT
Thousands of cancer patients are currently in clinical trials evaluating antiangiogenic therapy in the neoadjuvant setting, which is the treatment of localized primary tumors prior to surgical intervention. The rationale is that shrinking a tumor will improve surgical outcomes and minimize growth of occult micrometastatic disease-thus delaying post-surgical recurrence and improving survival. But approved VEGF pathway inhibitors have not been tested in clinically relevant neoadjuvant models that compare pre- and post-surgical treatment effects. Using mouse models of breast, kidney, and melanoma metastasis, we demonstrate that primary tumor responses to neoadjuvant VEGFR TKI treatment do not consistently correlate with improved post-surgical survival, with survival worsened in certain settings. Similar negative effects did not extend to protein-based VEGF pathway inhibitors and could be reversed with altered dose, surgical timing, and treatment duration, or when VEGFR TKIs are combined with metronomic 'anti-metastatic' chemotherapy regimens. These studies represent the first attempt to recapitulate the complex clinical parameters of neoadjuvant therapy in mice and identify a novel tool to compare systemic antiangiogenic treatment effects on localized and disseminated disease.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Supelco
5-Fluorouracil, analytical standard
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sunitinib malate, ≥98% (HPLC)