Merck
  • A multi-analyte LC-MS/MS method for the analysis of 23 mycotoxins in different sorghum varieties: the forgotten sample matrix.
A multi-analyte LC-MS/MS method for the analysis of 23 mycotoxins in different sorghum varieties: the forgotten sample matrix.
Food chemistry (2015-02-11)
Emmanuel Njumbe Ediage, Christof Van Poucke, Sarah De Saeger
ABSTRACT
An LC-MS/MS method was developed and validated for the detection and quantification of 23 mycotoxins in different varieties of sorghum. The method performance characteristics were as follows: suitable linearity ranges for all 23 mycotoxins with p-value >0.05; limits of detection (1.2-50 μg/kg), limits of quantification (2.5-100 μg/kg), repeatability (RSDr, 7-22%), intermediate precision (RSDR, 14-44%) and apparent recovery (0.2-11%, expressed as bias). The method was applied to analyze 10 samples obtained from retail shops in Belgium (n=8) and Germany (n=2). Nine of the 10 samples (90%) were positive for the following mycotoxins: aflatoxin B1 (50 μg/kg), alternariol monomethyl ether (<LOQ - 79 μg/kg), alternariol (303-357 μg/kg), diacetoxyscirpenol (<LOQ - 91 μg/kg), fumonisin B1 (<LOQ - 95 μg/kg), fumonisin B2 (<LOQ), fumonisin B3 (<LOQ), T2 (<LOQ) and zearalenone (<LOQ).
Product Description
Aflatoxin B2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Supelco
15-Acetyldeoxynivalenol solution, ~100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Methanol, SAJ special grade
Supelco
3-Acetyldeoxynivalenol solution, ~100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Fumonisin B1, reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
3-Acetyldeoxynivalenol
Supelco
Aflatoxin B2
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Supelco
Fumonisin B1 solution, ~50 μg/mL in acetonitrile: water, analytical standard
Supelco
Aflatoxin B2 solution, 0.5 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
3-Acetyldeoxynivalenol, from Fusarium roseum
Sigma-Aldrich
15-O-Acetyl-4-deoxynivalenol from Fusarium graminearum
Sigma-Aldrich
Aflatoxin B2
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Sigma-Aldrich
Zearalanone
Supelco
Sterigmatocystin
Sigma-Aldrich
Sterigmatocystin, powder, ≥98% (TLC)
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG