Merck
  • Dairy proteins, dairy lipids, and postprandial lipemia in persons with abdominal obesity (DairyHealth): a 12-wk, randomized, parallel-controlled, double-blinded, diet intervention study.
Dairy proteins, dairy lipids, and postprandial lipemia in persons with abdominal obesity (DairyHealth): a 12-wk, randomized, parallel-controlled, double-blinded, diet intervention study.
The American journal of clinical nutrition (2015-04-04)
Mette Bohl, Ann Bjørnshave, Kia V Rasmussen, Anne Grethe Schioldan, Bashar Amer, Mette K Larsen, Trine K Dalsgaard, Jens J Holst, Annkatrin Herrmann, Sadhbh O'Neill, Lorraine O'Driscoll, Lydia Afman, Erik Jensen, Merete M Christensen, Søren Gregersen, Kjeld Hermansen
ABSTRACT
Abdominal obesity and exaggerated postprandial lipemia are independent risk factors for cardiovascular disease (CVD) and mortality, and both are affected by dietary behavior. We investigated whether dietary supplementation with whey protein and medium-chain saturated fatty acids (MC-SFAs) improved postprandial lipid metabolism in humans with abdominal obesity. We conducted a 12-wk, randomized, double-blinded, diet intervention study. Sixty-three adults were randomly allocated to one of 4 diets in a 2 × 2 factorial design. Participants consumed 60 g milk protein (whey or casein) and 63 g milk fat (with high or low MC-SFA content) daily. Before and after the intervention, a high-fat meal test was performed. We measured changes from baseline in fasting and postprandial triacylglycerol, apolipoprotein B-48 (apoB-48; reflecting chylomicrons of intestinal origin), free fatty acids (FFAs), insulin, glucose, glucagon, glucagon-like peptide 1 (GLP-1), and gastric inhibitory polypeptide (GIP). Furthermore, changes in the expression of adipose tissue genes involved in lipid metabolism were investigated. Two-factor ANOVA was used to examine the difference between protein types and fatty acid compositions, as well as any interaction between the two. Fifty-two participants completed the study. We found that the postprandial apoB-48 response decreased significantly after whey compared with casein (P = 0.025) independently of fatty acid composition. Furthermore, supplementation with casein resulted in a significant increase in the postprandial GLP-1 response compared with whey (P = 0.003). We found no difference in postprandial triacylglycerol, FFA, insulin, glucose, glucagon, or GIP related to protein type or MC-SFA content. We observed no interaction between milk protein and milk fat on postprandial lipemia. We found that a whey protein supplement decreased the postprandial chylomicron response compared with casein in persons with abdominal obesity, thereby indicating a beneficial impact on CVD risk. This trial was registered at clinicaltrials.gov as NCT01472666.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Nitrogen, ≥99.998%
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material