Merck
  • Selective removal of phosphate for analysis of organic acids in complex samples.
Selective removal of phosphate for analysis of organic acids in complex samples.
Journal of chromatography. A (2015-03-01)
Sandeep Deshmukh, Andrej Frolov, Andrea Marcillo, Claudia Birkemeyer
ABSTRACT
Accurate quantitation of compounds in samples of biological origin is often hampered by matrix interferences one of which occurs in GC-MS analysis from the presence of highly abundant phosphate. Consequently, high concentrations of phosphate need to be removed before sample analysis. Within this context, we screened 17 anion exchange solid-phase extraction (SPE) materials for selective phosphate removal using different protocols to meet the challenge of simultaneous recovery of six common organic acids in aqueous samples prior to derivatization for GC-MS analysis. Up to 75% recovery was achieved for the most organic acids, only the low pKa tartaric and citric acids were badly recovered. Compared to the traditional approach of phosphate removal by precipitation, SPE had a broader compatibility with common detection methods and performed more selectively among the organic acids under investigation. Based on the results of this study, it is recommended that phosphate removal strategies during the analysis of biologically relevant small molecular weight organic acids consider the respective pKa of the anticipated analytes and the detection method of choice.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
D-(−)-Fructose, SAJ special grade, ≥98.0%
Sigma-Aldrich
D-Mannitol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Sigma-Aldrich
D-Mannitol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Adonitol, BioXtra, ≥99.0% (HPLC)
Supelco
Methanol, analytical standard
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)