Merck
  • Fibrosis, vascular activation, and immune abnormalities resembling systemic sclerosis in bleomycin-treated Fli-1-haploinsufficient mice.
Fibrosis, vascular activation, and immune abnormalities resembling systemic sclerosis in bleomycin-treated Fli-1-haploinsufficient mice.
Arthritis & rheumatology (Hoboken, N.J.) (2014-11-12)
Takashi Taniguchi, Yoshihide Asano, Kaname Akamata, Shinji Noda, Takehiro Takahashi, Yohei Ichimura, Tetsuo Toyama, Maria Trojanowska, Shinichi Sato
ABSTRACT
Fli-1, a potential predisposing factor for systemic sclerosis (SSc), is constitutively down-regulated in the lesional skin of patients with SSc by an epigenetic mechanism. To investigate the impact of Fli-1 deficiency on the induction of an SSc phenotype in various cell types, we generated bleomycin-induced skin fibrosis in Fli-1(+/-) mice and investigated the molecular mechanisms underlying its phenotypic alterations. Messenger RNA (mRNA) levels and protein expression of target molecules were examined by quantitative reverse transcription-polymerase chain reaction and immunostaining. Transforming growth factor β (TGFβ) bioassay was used to evaluate the activation of latent TGFβ. The binding of Fli-1 to the target gene promoters was assessed with chromatin immunoprecipitation. Bleomycin induced more severe dermal fibrosis in Fli-1(+/-) mice than in wild-type mice. Fli-1 haploinsufficiency activated dermal fibroblasts via the up-regulation of αvβ3 and αvβ5 integrins and activation of latent TGFβ. Dermal fibrosis in Fli-1(+/-) mice was also attributable to endothelial-to-mesenchymal transition, which is directly induced by Fli-1 deficiency and amplified by bleomycin. Th2/Th17-skewed inflammation and increased infiltration of mast cells and macrophages were seen, partly due to the altered expression of cell adhesion molecules in endothelial cells as well as the induction of the skin chemokines. Fli-1(+/-) mouse macrophages preferentially differentiated into an M2 phenotype upon stimulation with interleukin-4 (IL-4) or IL-13. Our findings provide strong evidence for the fundamental role of Fli-1 deficiency in inducing SSc-like phenotypic alterations in dermal fibroblasts, endothelial cells, and macrophages in a manner consistent with human disease.
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard