Merck
  • Lattice water molecules tuned spin-crossover for an iron(II) complex with thermal hysteresis.
Lattice water molecules tuned spin-crossover for an iron(II) complex with thermal hysteresis.
Dalton transactions (Cambridge, England : 2003) (2014-10-11)
Yang-Hui Luo, Li-Jing Yang, Qing-Ling Liu, Yang Ling, Wei Wang, Bai-Wang Sun
ABSTRACT
A new iron(II) complex based on the 4,4'-dimethyl-2,2'-bipyridine ligand [Fe(4,4'-dmbpy)3(ClO4)(SCN)·3H2O (1·3H2O)] has been prepared and characterized. Structural studies and Hirshfeld surface analysis for complex 1·3H2O at three different temperatures (300, 240 and 130 K) are described. The UV-vis absorption spectrum of a water-free sample (1) in methanol solution and magnetic susceptibility measurements for solid-state samples 1·3H2O and 1 revealed that the removal of lattice water molecules from complex 1·3H2O changed the magnetic properties from the low-spin state (1·3H2O) to the complete spin-crossover (1) between 350-220 K with a thermal hysteresis of 7 K, and was accompanied by a colour change from brown to red.
Sigma-Aldrich
Potassium thiocyanate, JIS special grade, ≥99.5%
Sigma-Aldrich
4,4′-Dimethyl-2,2′-dipyridyl, 99%
Sigma-Aldrich
4,4′-Dimethyl-2,2′-dipyridyl, 99.5%, purified by sublimation
Sigma-Aldrich
Potassium thiocyanate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium thiocyanate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium thiocyanate, ACS reagent, 99%
Sigma-Aldrich
Potassium thiocyanate solution, 0.1 M
Sigma-Aldrich
Potassium thiocyanate solution, volumetric, 8 M KSCN