Merck
  • Expeditious low-temperature sintering of copper nanoparticles with thin defective carbon shells.
Expeditious low-temperature sintering of copper nanoparticles with thin defective carbon shells.
Nanoscale (2015-03-21)
Changkyu Kim, Gyoungja Lee, Changkyu Rhee, Minku Lee
ABSTRACT
The realization of air-stable nanoparticles, well-formulated nanoinks, and conductive patterns based on copper is a great challenge in low-cost and large-area flexible printed electronics. This work reports the synthesis of a conductively interconnected copper structure via thermal sintering of copper inks at a low temperature for a short period of time, with the help of thin defective carbon shells coated onto the copper nanoparticles. Air-stable copper/carbon core/shell nanoparticles (typical size ∼23 nm, shell thickness ∼1.0 nm) are prepared by means of an electric explosion of wires. Gaseous oxidation of the carbon shells with a defective structure occurs at 180 °C, impacting the choice of organic solvents as well as the sintering conditions to create a crucial neck formation. Isothermal oxidation and reduction treatment at 200 °C for only about 10 min yields an oxide-free copper network structure with an electrical resistivity of 25.1 μΩ cm (14.0 μΩ cm at 250 °C). Finally, conductive copper line patterns are achieved down to a 50 μm width with an excellent printing resolution (standard deviation ∼4.0%) onto a polyimide substrate using screen printing of the optimized inks.
Sigma-Aldrich
Dicyandiamide, 99%
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
USP
Metformin Related Compound A, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Metformin impurity A, European Pharmacopoeia (EP) Reference Standard
Supelco
Metformin Related Compound A, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
α-Terpineol, 90%, technical grade
Sigma-Aldrich
Terpineol, mixture of isomers, 96%, FG
Sigma-Aldrich
Di(ethylene glycol) ethyl ether, ≥99%
Supelco
Ethylene glycol butyl ether, analytical standard
Sigma-Aldrich
Diethylene glycol monoethyl ether, SAJ first grade, ≥98.0%
Sigma-Aldrich
Ethylene glycol butyl ether, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
USP
Diethylene glycol monoethyl ether, United States Pharmacopeia (USP) Reference Standard
Supelco
α-Terpineol, analytical standard
Sigma-Aldrich
Ethylene glycol butyl ether, spectrophotometric grade, ≥99.0%
Sigma-Aldrich
Ethylene glycol butyl ether, ≥99%
Sigma-Aldrich
Diethylene glycol monoethyl ether, ReagentPlus®, 99%