Merck
  • Intravenous infusion of electrolyte solution changes pharmacokinetics of drugs: pharmacokinetics of ampicillin.
Intravenous infusion of electrolyte solution changes pharmacokinetics of drugs: pharmacokinetics of ampicillin.
Journal of veterinary pharmacology and therapeutics (2014-03-29)
M Britzi, Y Mazon, E Lavy, S Soback
ABSTRACT
The pharmacokinetics of ampicillin in dogs was determined after intravenous (i.v.) bolus and constant rate infusion. Ampicillin was administered to six beagle dogs as an i.v. bolus at 20 mg/kg and as a constant rate i.v. infusion (CRI) at 20 mg/kg during 8 h (0.042 mL/min/kg) in Ringer's lactate (Hartmann's) solution. The concentrations were determined by an LC/MS/MS method. After i.v. bolus, ampicillin total body clearance, apparent volume of distribution at steady-state, mean residence time (MRT), and half-life were 4.53 ± 0.70 mL/min/kg, 0.275 ± 0.044 L/kg, 61 ± 13 min, and 111 (85-169) min, respectively. The corresponding parameters calculated after CRI were 13.5 ± 1.06 mL/min/kg, 0.993 ± 0.415 L/kg, 73 ± 27 min, and 49 (31-69) min. Ampicillin concentration decreased by 30% in the Ringer's lactate infusion solution mostly during the first hour after preparation of the solution. Constant rate infusion of Ringer's lactate solution during 8 h caused significant changes in ampicillin pharmacokinetics. The results suggested that special attention should be given to drug pharmacokinetics when co-administered intravenously with electrolyte solutions.
Product Description
Supelco
Ampicillin trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Cloxacillin sodium, European Pharmacopoeia (EP) Reference Standard
Ampicillin trihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cloxacillin sodium salt, ≥95.0% (HPLC)
Sigma-Aldrich
Ampicillin trihydrate, 900-1050 μg/mg anhydrous basis (HPLC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
USP
Ampicillin trihydrate, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cloxacillin sodium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Ampicillin Trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%