Merck
  • The effects of C-glycosylation of luteolin on its antioxidant, anti-Alzheimer's disease, anti-diabetic, and anti-inflammatory activities.
The effects of C-glycosylation of luteolin on its antioxidant, anti-Alzheimer's disease, anti-diabetic, and anti-inflammatory activities.
Archives of pharmacal research (2014-07-06)
Jae Sue Choi, Md Nurul Islam, Md Yousof Ali, Young Myeong Kim, Hye Jin Park, Hee Sook Sohn, Hyun Ah Jung
ABSTRACT
To investigate the effect of C-glycosylation at different positions of luteolin, the structure-activity relationships of luteolin and a pair of isomeric C-glycosylated derivatives orientin and isoorientin, were evaluated. We investigated the effects of C-glycosylation on the antioxidant, anti-Alzheimer's disease (AD), anti-diabetic and anti-inflammatory effects of luteolin and its two C-glycosides via in vitro assays of peroxynitrite (ONOO(-)), total reactive oxygen species (ROS), nitric oxide (NO), 1,1-diphenyl-2-picrylhydraxyl (DPPH), aldose reductase, protein tyrosine phosphatase 1B (PTP1B), acetylcholinesterase (AChE), butyrylcholinesterase (BChE), and β-site amyloid precursor cleaving enzyme 1 (BACE1), and cellular assays of NO production and inducible nitric oxide synthase (iNOS)/cyclooxygenase-2 expression in lipopolysaccharide (LPS)-stimulated RAW 264.7 cells. Of the three compounds, isoorientin showed the highest scavenging activity against DPPH, NO, and ONOO(-), while luteolin was the most potent inhibitor of ROS generation. In addition, luteolin showed the most potent anti-AD activity as determined by its inhibition of AChE, BChE, and BACE1. With respect to anti-diabetic effects, luteolin exerted the strongest inhibitory activity against PTP1B and rat lens aldose reductase. Luteolin also inhibited NO production and iNOS protein expression in LPS-stimulated macrophages, while orientin and isoorientin were inactive at the same concentrations. The effects of C-glycosylation at different positions of luteolin may be closely linked to the intensity and modulation of antioxidant, anti-AD, anti-diabetic, and anti-inflammatory effects of luteolin and its C-glycosylated derivatives.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
DL-Glyceraldehyde, dimer, 95%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, for complexometry, ≥99.0%
Sigma-Aldrich
L-Penicillamine, 99%
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
NGF-β human, from human, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline