Merck
  • Vitamin D reverses aPL-induced inflammation and LMWH-induced sFlt-1 release by human trophoblast.
Vitamin D reverses aPL-induced inflammation and LMWH-induced sFlt-1 release by human trophoblast.
American journal of reproductive immunology (New York, N.Y. : 1989) (2014-07-30)
Stefan M Gysler, Melissa J Mulla, Meredith Stuhlman, Anna K Sfakianaki, Michael J Paidas, Nancy L Stanwood, Aileen Gariepy, Jan J Brosens, Lawrence W Chamley, Vikki M Abrahams
ABSTRACT
Women with antiphospholipid syndrome (APS) are at increased risk of recurrent pregnancy loss (RPL) and preeclampsia. Antiphospholipid antibodies (aPL) directly alter trophoblast function. Treatment with low molecular weight heparin (LMWH) reduces the risk of RPL but not preeclampsia. Moreover, LMWH stimulates trophoblast sFlt-1 release, an anti-angiogenic factor associated with preeclampsia. Since vitamin D deficiency is associated with APS and preeclampsia, this study sought to determine the effect of vitamin D on trophoblast function in the setting of aPL and LMWH. A human first trimester trophoblast cell line (HTR8) and primary trophoblast cultures were treated with or without aPL in the presence and absence of vitamin D, LMWH or both. Trophoblast secretion of inflammatory cytokines and angiogenic factors were measured by ELISA. Vitamin D alone or in combination with LMWH attenuated the aPL-induced trophoblast inflammatory response in the HTR8 cells and primary cultures. While vitamin D did not have any impact on aPL-mediated modulation of angiogenic factors in the primary trophoblast, it significantly inhibited LMWH-induced sFlt-1 release. LMWH in combination with vitamin D may be more beneficial than single-agent therapy by preventing aPL-induced trophoblast inflammation and reversing LMWH-induced sFlt-1 secretion.
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
D-Valine, suitable for cell culture, BioReagent
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
D-Valine, ≥98%
Tyrosine, European Pharmacopoeia (EP) Reference Standard