Merck
  • 3D artificial bones for bone repair prepared by computed tomography-guided fused deposition modeling for bone repair.
3D artificial bones for bone repair prepared by computed tomography-guided fused deposition modeling for bone repair.
ACS applied materials & interfaces (2014-08-19)
Ning Xu, Xiaojian Ye, Daixu Wei, Jian Zhong, Yuyun Chen, Guohua Xu, Dannong He
ABSTRACT
The medical community has expressed significant interest in the development of new types of artificial bones that mimic natural bones. In this study, computed tomography (CT)-guided fused deposition modeling (FDM) was employed to fabricate polycaprolactone (PCL)/hydroxyapatite (HA) and PCL 3D artificial bones to mimic natural goat femurs. The in vitro mechanical properties, in vitro cell biocompatibility, and in vivo performance of the artificial bones in a long load-bearing goat femur bone segmental defect model were studied. All of the results indicate that CT-guided FDM is a simple, convenient, relatively low-cost method that is suitable for fabricating natural bonelike artificial bones. Moreover, PCL/HA 3D artificial bones prepared by CT-guided FDM have more close mechanics to natural bone, good in vitro cell biocompatibility, biodegradation ability, and appropriate in vivo new bone formation ability. Therefore, PCL/HA 3D artificial bones could be potentially be of use in the treatment of patients with clinical bone defects.
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Methyl methacrylate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Methyl methacrylate, European Pharmacopoeia (EP) Reference Standard