Merck
  • Recombinant human IgA1 and IgA2 autoantibodies to type VII collagen induce subepidermal blistering ex vivo.
Recombinant human IgA1 and IgA2 autoantibodies to type VII collagen induce subepidermal blistering ex vivo.
Journal of immunology (Baltimore, Md. : 1950) (2014-07-16)
Andreas Recke, Luisa M Trog, Hendri H Pas, Artem Vorobyev, Aida Abadpour, Marcel F Jonkman, Ger van Zandbergen, Claudia Kauderer, Detlef Zillikens, Gestur Vidarsson, Ralf J Ludwig
ABSTRACT
Subepidermal autoimmune blistering dermatoses (AIBD) are prototypic autoantibody-mediated diseases. In epidermolysis bullosa acquisita (EBA), an autoimmune disease with severe and chronic skin blistering, autoantibodies are directed against type VII collagen. IgG is the predominant autoantibody isotype of EBA, the pathogenicity of which has been demonstrated in a variety of in vivo and ex vivo disease models. In contrast, there is not much evidence for the pathogenicity of IgA, which may appear as the only autoantibody isotype in some EBA patients. To investigate the pathogenic potential of IgA autoantibodies, we generated chimeric V gene-matched human IgA1, IgA2, and control IgG1 autoantibodies directed against type VII collagen. Immobilized immune complexes containing the rIgA1 and rIgA2 autoantibodies induced the dose-dependent release of reactive oxygen species from neutrophil granulocytes, a precondition for blister formation. Moreover, both rIgA1 and rIgA2 induced leukocyte-dependent dermal-epidermal separation in cryosections of human skin. In contrast with rIgG1, neither rIgA1 nor rIgA2 was capable of inducing complement deposition at the dermal-epidermal junction. Because complement activation is a prerequisite for blister induction, this lack of function compared with IgG1 may be compensated for by the stronger activation of neutrophil granulocytes by both IgA1 and IgA2. For IgG-mediated AIBD, immunoadsorption therapy is a convenient treatment modality for the removal of pathogenic autoantibodies, particularly in treatment-resistant cases. The results of this study show the pathogenic potential of IgA autoantibodies and support the development of adsorber matrices for IgA-mediated AIBD.
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Barbital, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Barbital
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Barbital, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Anti-Human IgG1−FITC antibody, Mouse monoclonal, clone 8c/6-39, purified from hybridoma cell culture