Merck
  • Comparative effect of telmisartan vs lisinopril on blood pressure in patients of metabolic syndrome.
Comparative effect of telmisartan vs lisinopril on blood pressure in patients of metabolic syndrome.
Endocrine, metabolic & immune disorders drug targets (2014-12-03)
Pradip N Gore, Vandana A Badar, Mrunalini M Hardas, Varsha J Bansode
ABSTRACT
The present study was planned to focus on comparative effects of telmisartan vs lisinopril on blood pressure in patients of metabolic syndrome The study was carried out on 62 patients of metabolic syndrome from Dec 2010 to Oct 2012 in OPD of Institute. There were two groups, A and B. Group A- Telmisartan (31 patients) and Group B- Lisinopril (31 patients) receiving Telmisartan 40 mg and lisinopril 5 mg orally once a day respectively for 12 weeks. The diagnosis of essential hypertension was made by the physician based on two measurements of blood pressure on two different occasions using auscultatory method and was done at initial stage and repeated after 6 weeks and 12 weeks of treatment in Group A and Group B patients. Our study found that telmisartan or lisinopril treatment for 12 weeks leads to statistically significant (p<0.001) reduction in both SBP and DBP at 6 and 12 weeks when compared with baseline, whereas comparison between telmisartan and lisinopril treatment failed to show any statistically significant effect. Treatment of metabolic patients with telmisartan or lisinopril for the management of hypertension reduced both Systolic blood pressure (SBP) as well as Diastolic blood pressure (DBP) statistically significantly during 12 weeks treatment. However, telmisartan and lisinopril treatment were found effective.
Product Description
Telmisartan for peak identification, European Pharmacopoeia (EP) Reference Standard
Telmisartan for system suitability, European Pharmacopoeia (EP) Reference Standard
Telmisartan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lisinopril, ≥98% (HPLC)
Sigma-Aldrich
Telmisartan, ≥98% (HPLC), solid
Supelco
Lisinopril, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Telmisartan, United States Pharmacopeia (USP) Reference Standard
USP
Lisinopril, United States Pharmacopeia (USP) Reference Standard