Merck
Effects of baicalin against UVA-induced photoaging in skin fibroblasts.
The American journal of Chinese medicine (2014-05-30)
Wei Min, Xin Liu, Qihong Qian, Bingjiang Lin, Di Wu, Miaomiao Wang, Israr Ahmad, Nabiha Yusuf, Dan Luo
ABSTRACT
Ultraviolet A (UVA) radiation contributes to skin photoaging. Baicalin, a plant-derived flavonoid, effectively absorbs UV rays and has been shown to have anti-oxidant and anti-inflammatory properties that may delay the photoaging process. In the current study, cultured human skin fibroblasts were incubated with 50 μg/ml baicalin 24 hours prior to 10 J/cm(2) UVA irradiation. In order to examine the efficacy of baicalin treatment in delaying UVA-induced photoaging, we investigated aging-related markers, cell cycle changes, anti-oxidant activity, telomere length, and DNA damage markers. UVA radiation caused an increased proportion of β-Gal positive cells and reduced telomere length in human skin fibroblasts. In addition, UVA radiation inhibited TGF-β1 secretion, induced G1 phase arrest, reduced SOD and GSH-Px levels, increased MDA levels, enhanced the expression of MMP-1, TIMP-1, p66, p53, and p16 mRNA, reduced c-myc mRNA expression, elevated p53 and p16 protein expression, and reduced c-myc protein expression. Baicalin treatment effectively protected human fibroblasts from these UVA radiation-induced aging responses, suggesting that the underlying mechanism involves the inhibition of oxidative damage and regulation of the expression of senescence-related genes, including those encoding for p53, p66(Shc) and p16.
Product Description
Baicalin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-TP53 (ab3) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Baicalin, 95%