Merck
  • A simple LC-MS/MS method using HILIC chromatography for the determination of fosfomycin in plasma and urine: application to a pilot pharmacokinetic study in humans.
A simple LC-MS/MS method using HILIC chromatography for the determination of fosfomycin in plasma and urine: application to a pilot pharmacokinetic study in humans.
Journal of pharmaceutical and biomedical analysis (2014-12-22)
Suzanne L Parker, Jeffrey Lipman, Jason A Roberts, Steven C Wallis
ABSTRACT
A high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) method, using hydrophilic interaction liquid chromatography (HILIC) chromatography for the analysis of fosfomycin in human plasma and urine, has been developed and validated. The plasma method uses a simple protein precipitation using a low volume sample (10 μL) and is suitable for the concentration range of 1 to 2000 μg/mL. The urine method involves a simple dilution of 10 μL of sample and is suitable for a concentration range of 0.1 to 10 mg/mL. The plasma and urine results, reported, respectively, are for recovery (68, 72%), inter-assay precision (≤9.1%, ≤8.1%) and accuracy (range -7.2 to 3.3%, -1.9 to 1.6%), LLOQ precision (4.7%, 3.1%) and accuracy (1.7% and 1.2%), and includes investigations into the linearity, stability and matrix effects. The method was used in a pilot pharmacokinetic study of a critically ill patient receiving i.v. fosfomycin, which measured a maximum and minimum plasma concentration of 222 μg/mL and 172 μg/mL, respectively, after the initial dose, and a maximum and minimum plasma concentration of 868 μg/mL and 591μg/mL, respectively, after the fifth dose. The urine concentration was 2.03 mg/mL after the initial dose and 0.29 mg/mL after the fifth dose.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), for copper determination
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ethylphosphonic acid, 98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%