Merck
  • Antibacterial, antifungal, phytotoxic, and genotoxic properties of two complexes of Ag(I) with sulfachloropyridazine (SCP): X-ray diffraction of [Ag(SCP)]n.
Antibacterial, antifungal, phytotoxic, and genotoxic properties of two complexes of Ag(I) with sulfachloropyridazine (SCP): X-ray diffraction of [Ag(SCP)]n.
ChemMedChem (2014-05-09)
Natalia Mosconi, Cecilia Giulidori, Francesca Velluti, Estela Hure, Agustina Postigo, Graciela Borthagaray, Davi Fernando Back, María H Torre, Marcela Rizzotto
ABSTRACT
We report the synthesis, characterization, antibacterial and antifungal activities, phytotoxicity, and genotoxicity of two new complexes of silver(I) with sulfachloropyridazine (SCP), one of which is heteroleptic with SCP and SCN(-) ligands (Ag-SCP-SCN), the other of which is homoleptic (Ag-SCP); furthermore, the crystal structure of the homoleptic complex is disclosed. The heterocyclic N atom nearest to the Cl atom and the N(sulfonamide) atom could be coordination sites for the silver ion in the Ag-SCP-SCN complex. The Ag-SCP complex is a polymeric compound with metal-metal bonds, and the heterocyclic and sulfonamide N atoms are points of coordination for Ag(I) . Both complexes showed activity against all the tested bacteria, and in the cases of Escherichia coli and Pseudomonas aeruginosa, the action was better than that of SCP. In all cases, both silver-SCP complexes showed better antifungal activity than SCP, which was inactive against the tested fungi. Notably, the activity against P. aeruginosa, a nosocomial multidrug-resistant pathogen, was better than that of the reference antibiotic cefotaxim. Both silver-sulfa complexes displayed moderate activity against the tested yeast, especially for C. neoformans, which is an important fact considering the incidence of cryptococcosis, mainly in immune-deficient patients. No chromosomal aberrations were observed with the Allium cepa test, which is auspicious for further study of these complexes as potential drugs.
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
Sigma-Aldrich
Sodium metabisulfite, JIS first grade, ≥95.0%
Sigma-Aldrich
Sodium metabisulfite, ReagentPlus®, ≥99%
Supelco
Sodium Metabisulfite, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium metabisulfite, reagent grade, 97%
Sigma-Aldrich
Sodium metabisulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Supelco
Sodium metabisulfite, analytical standard
Sigma-Aldrich
Sodium metabisulfite, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium metabisulfite, puriss. p.a., ACS reagent, reag. Ph. Eur., dry, 98-100.5%
USP
Sodium metabisulfite, United States Pharmacopeia (USP) Reference Standard