Merck
Artifact produced in disc electrophoresis by ammonium persulfate.
Science (New York, N.Y.) (1967-04-14)
J M Brewer
ABSTRACT
Ammonium persulfate, a common polymerizing agent for acrylamide gels, can inactivate yeast enolase and produce increased electrophoretic heterogeneity during disc electrophoresis in gels containing 8M urea. The use of riboflavin and light for polymerization or thioglycolate for removal of the persulfate are feasible alternatives.
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%