Merck
A protective chimeric antibody to tick-borne encephalitis virus.
Vaccine (2014-05-20)
Ivan K Baykov, Andrey L Matveev, Oleg V Stronin, Alexander B Ryzhikov, Leonid E Matveev, Marat F Kasakin, Vladimir A Richter, Nina V Tikunova
ABSTRACT
The efficiency of several mouse monoclonal antibodies (mAbs) specific to the tick-borne encephalitis virus (TBEV) glycoprotein E in post-exposure prophylaxis was assessed, and mAb14D5 was shown to be the most active of all those studied. It was proven that the hybridoma cell line 14D5 produced one immunoglobulin H chain and two L chains. They were used to construct chimeric antibodies ch14D5a and ch14D5b, the affinity constants of which were 2.6 × 10(10)M(-1) and 1.0 × 10(7)M(-1), respectively, according to the SPR-based ProteOn biosensor assay. The neutralization index (IC50) of ch14D5a was 0.04 μg/ml in the focus reduction neutralization test. In in vivo experiments, ch14D5a at a dose of 10 μg/mouse resulted in a 100% survival of the mice infected with 240 LD50 of TBEV. This chimeric antibody is promising for further development of prevention and therapeutic drugs against TBEV.
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
trans-Sinapic acid, analytical standard
Supelco
2,5-Dihydroxybenzoic acid, analytical standard
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure