Merck
  • Integrated metabolic spatial-temporal model for the prediction of ammonia detoxification during liver damage and regeneration.
Integrated metabolic spatial-temporal model for the prediction of ammonia detoxification during liver damage and regeneration.
Hepatology (Baltimore, Md.) (2014-03-29)
Freimut Schliess, Stefan Hoehme, Sebastian G Henkel, Ahmed Ghallab, Dominik Driesch, Jan Böttger, Reinhard Guthke, Michael Pfaff, Jan G Hengstler, Rolf Gebhardt, Dieter Häussinger, Dirk Drasdo, Sebastian Zellmer
ABSTRACT
The impairment of hepatic metabolism due to liver injury has high systemic relevance. However, it is difficult to calculate the impairment of metabolic capacity from a specific pattern of liver damage with conventional techniques. We established an integrated metabolic spatial-temporal model (IM) using hepatic ammonia detoxification as a paradigm. First, a metabolic model (MM) based on mass balancing and mouse liver perfusion data was established to describe ammonia detoxification and its zonation. Next, the MM was combined with a spatial-temporal model simulating liver tissue damage and regeneration after CCl4 intoxication. The resulting IM simulated and visualized whether, where, and to what extent liver damage compromised ammonia detoxification. It allowed us to enter the extent and spatial patterns of liver damage and then calculate the outflow concentrations of ammonia, glutamine, and urea in the hepatic vein. The model was validated through comparisons with (1) published data for isolated, perfused livers with and without CCl4 intoxication and (2) a set of in vivo experiments. Using the experimentally determined portal concentrations of ammonia, the model adequately predicted metabolite concentrations over time in the hepatic vein during toxin-induced liver damage and regeneration in rodents. Further simulations, especially in combination with a simplified model of blood circulation with three ammonia-detoxifying compartments, indicated a yet unidentified process of ammonia consumption during liver regeneration and revealed unexpected concomitant changes in amino acid metabolism in the liver and at extrahepatic sites. The IM of hepatic ammonia detoxification considerably improves our understanding of the metabolic impact of liver disease and highlights the importance of integrated modeling approaches on the way toward virtual organisms.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Urea, analytical standard