Merck
  • The response of amphibian larvae to exposure to a glyphosate-based herbicide (Roundup WeatherMax) and nutrient enrichment in an ecosystem experiment.
The response of amphibian larvae to exposure to a glyphosate-based herbicide (Roundup WeatherMax) and nutrient enrichment in an ecosystem experiment.
Ecotoxicology and environmental safety (2014-09-01)
Christopher Edge, Dean Thompson, Chunyan Hao, Jeff Houlahan
ABSTRACT
Herbicides and fertilizers are widely used throughout the world and pose a threat to aquatic ecosystems. Using a replicated, whole ecosystem experiment in which 24 small wetlands were split in half with an impermeable barrier we tested whether exposure to a glyphosate-based herbicide, Roundup WeatherMax™, alone or in combination with nutrient enrichment has an effect on the survival, growth or development of amphibians. The herbicide was applied at one of two concentrations (low=210 μg a.e./L, high=2880 μg a.e./L) alone and in combination with nutrient enrichment to one side of wetlands and the other was left as an untreated control. Each treatment was replicated with six wetlands, and the experiment was repeated over two years. In the high glyphosate and nutrient enrichment treatment the survival of wood frog (Lithobates sylvaticus) larvae was lower in enclosures placed in situ on the treated sides than the control sides of wetlands. However, these results were not replicated in the second year of study and they were not observed in free swimming wood frog larvae in the wetlands. In all treatments, wood frog larvae on the treated sides of wetlands were slightly larger (<10%) than those on the control side, but no effect on development was observed. The most dramatic finding was that the abundance of green frog larvae (Lithobates clamitans) was higher on the treated sides than the control sides of wetlands in the herbicide and nutrient treatments during the second year of the study. The results observed in this field study indicate that caution is necessary when extrapolating results from artificial systems to predict effects in natural systems. In this experiment, the lack of toxicity to amphibian larvae was probably due to the fact the pH of the wetlands was relatively low and the presence of sediments and organic surfaces which would have mitigated the exposure duration.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SAFC
Glycine
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)