Merck
  • The uptake of ZnO and CuO nanoparticles in the water-flea Daphnia magna under acute exposure scenarios.
The uptake of ZnO and CuO nanoparticles in the water-flea Daphnia magna under acute exposure scenarios.
Environmental pollution (Barking, Essex : 1987) (2014-08-12)
Nathalie Adam, Frédéric Leroux, Dries Knapen, Sara Bals, Ronny Blust
ABSTRACT
In this study the uptake of ZnO and CuO nanoparticles by Daphnia magna was tested. Daphnids were exposed during 48 h to acute concentrations of the nanoparticles and corresponding metal salts. The Daphnia zinc and copper concentration was measured and the nanoparticles were localized using electron microscopy. The aggregation and dissolution in the medium was characterized. A fast dissolution of ZnO in the medium was observed, while most CuO formed large aggregates and only a small fraction dissolved. The Daphnia zinc concentration was comparable for the nanoparticles and salts. Contrarily, a much higher Daphnia copper concentration was observed in the CuO exposure, compared to the copper salt. CuO nanoparticles adsorbed onto the carapace and occurred in the gut but did not internalize in the tissues. The combined dissolution and uptake results indicate that the toxicity of both nanoparticle types was caused by metal ions dissolved from the particles in the medium.
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Copper, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Copper, tube, 1000mm, outside diameter 5.0mm, inside diameter 4mm, wall thickness 0.5mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.9%
Copper, rod, 50mm, diameter 6.35mm, as drawn, 99.999%
Copper, tube, 100mm, outside diameter 1.75mm, inside diameter 0.33mm, wall thickness 0.71mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 12.7mm, inside diameter 10.26mm, wall thickness 1.22mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 50mm disks, thickness 0.25mm, half hard, 99.95+%
Copper, wire reel, 100m, diameter 0.025mm, as drawn, 99.99+%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.7mm, half hard, 99.95+%
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Copper - O.F.H.C., foil, 50mm disks, thickness 0.5mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 6mm disks, thickness 0.006mm, hard, 99.95+%
Copper, wire reel, 100m, diameter 0.02mm, hard, 99.99+%
Copper - O.F.H.C., foil, 2m coil, thickness 0.125mm, hard, 99.95+%
Copper - O.F.H.C., foil, 25mm disks, thickness 1.0mm, half hard, 99.95+%
Copper, wire reel, 5m, diameter 0.5mm, as drawn, 99.98+%
Copper - O.F.H.C., foil, 2m coil, thickness 0.7mm, half hard, 99.95+%