Merck
  • Validation of an LC-ESI-MS/MS method for the quantitation of phosphodiesterase-5 inhibitors and their main metabolites in rat serum and brain tissue samples.
Validation of an LC-ESI-MS/MS method for the quantitation of phosphodiesterase-5 inhibitors and their main metabolites in rat serum and brain tissue samples.
Journal of pharmaceutical and biomedical analysis (2012-06-01)
Nora Unceta, Leire Echeazarra, Mario Montaña, Joan Sallés, Alberto Gómez-Caballero, M Aránzazu Goicolea, Ramón J Barrio
ABSTRACT
This work proposes a liquid chromatography-electrospray ionization ion trap mass spectrometry (LC-ESI-ITMS) method, for the quantification of sildenafil (SDF), tadalafil (TDF) and vardenafil (VDF) and their metabolites N-desmethylSDF, O-desethylSDF and N-desethylVDF, preceded by a sample preparation step based on protein and phospholipid elimination. A C8 column (150 mm × 4.6 mm, 5 μm) with ammonium formate (20mM) and acetonitrile as the mobile phase components have been used. This method has been validated, obtaining limits of quantification ranged from 1 to 2.5 ng/mL and 2 to 5 ng/g in serum and brain tissue respectively, while limits of detection ranged from 0.3 to 0.9 ng/mL in serum and 0.6 to 1.9 ng/g in brain tissue. Assay recoveries for low level QC samples were higher than 83% and the matrix effect ranged between 91% and 108% in serum and between 98% and 107% in brain tissue. The method has been applied to the quantification of these compounds in the serum and brain tissue of rats treated intraperitoneally with 10 mg/kg of SDF, TDF or VDF.
Product Description
Supelco
HybridSPE®-Phospholipid, Small Volume 96-well Plate, bed wt. 15 mg, volume 0.8 mL, pk of 1
Supelco
HybridSPE®-Phospholipid, Small Volume 96-well Plate, bed wt. 15 mg, volume 0.8 mL, pk of 20
Supelco
HybridSPE®-Phospholipid, Cartridge, bed wt. 30 mg, volume 1 mL, pk of 200, polypropylene material (hardware), PE frit (20 μm)
Supelco
HybridSPE®-Phospholipid, Cartridge, bed wt. 500 mg, volume 6 mL, pk of 30
Supelco
HybridSPE®-Phospholipid, Cartridge, bed wt. 30 mg, volume 1 mL, pk of 100
Supelco
HybridSPE®-Phospholipid, 96-well Plate, bed wt. 50 mg, volume 2 mL, pk of 1
Supelco
HybridSPE®-Phospholipid 96-Well Essentials Kit, 96-Well Essentials Kit
Supelco
HybridSPE®-Phospholipid Ultra, cartridge, bed wt. 30 mg, volume 1 mL, pk of 100