Merck
  • Specific interactions between Smad proteins and AP-1 components determine TGFβ-induced breast cancer cell invasion.
Specific interactions between Smad proteins and AP-1 components determine TGFβ-induced breast cancer cell invasion.
Oncogene (2012-08-29)
A Sundqvist, A Zieba, E Vasilaki, C Herrera Hidalgo, O Söderberg, D Koinuma, K Miyazono, C-H Heldin, U Landegren, P Ten Dijke, H van Dam
ABSTRACT
Deregulation of the transforming growth factor β (TGFβ) signal transduction cascade is functionally linked to cancer. In early phases, TGFβ acts as a tumor suppressor by inhibiting tumor cell proliferation, whereas in late phases, it can act as a tumor promoter by stimulating tumor cell invasion and metastasis. Smad transcriptional effectors mediate TGFβ responses, but relatively little is known about the Smad-containing complexes that are important for epithelial-mesenchymal transition and invasion. In this study, we have tested the hypothesis that specific members of the AP-1 transcription factor family determine TGFβ signaling specificity in breast cancer cell invasion. Using a 3D model of collagen-embedded spheroids of MCF10A-MII premalignant human breast cancer cells, we identified the AP-1 transcription factor components c-Jun, JunB, c-Fos and Fra1 as essential factors for TGFβ-induced invasion and found that various mesenchymal and invasion-associated TGFβ-induced genes are co-regulated by these proteins. In situ proximity ligation assays showed that TGFβ signaling not only induces complexes between Smad3 and Smad4 in the nucleus but also complexes between Smad2/3 and Fra1, whereas complexes between Smad3, c-Jun and JunB could already be detected before TGFβ stimulation. Finally, chromatin immunoprecipitations showed that c-Jun, JunB and Fra1, but not c-Fos, are required for TGFβ-induced binding of Smad2/3 to the mmp-10 and pai-1 promoters. Together these results suggest that in particular formation of Smad2/3-Fra1 complexes may reflect activation of the Smad/AP-1-dependent TGFβ-induced invasion program.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Nunc® Lab-Tek® II - CC2 Chamber Slide system, 8 wells, glass slide, 0.7 cm2/well, sterile
SAFC
Glycine
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 70 purifications
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Corning® Costar® TC-Treated Multiple Well Plates, size 6 wells, clear, polystyrene plate, flat bottom, case of 50 (individually wrapped), sterile, lid
Corning® 96 Well Clear Polystyrene Microplate, flat bottom clear, polystyrene, sterile, lid, case of 100, pack of 20
Corning® 96 Well Clear Polystyrene Microplate, round bottom clear, polystyrene, bag of 25, sterile, lid: no
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from salmon testes
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Laemmli Lysis-buffer, non smelling
Sigma-Aldrich
Tris buffered saline with Tween® 20, BioUltra, tablet (for 500 mL), pH 7.6
Sigma-Aldrich
Collagen solution from bovine skin, Type I, 2.9-3.2 mg/mL, suitable for cell culture, sterile-filtered
Sigma-Aldrich
GenElute PCR Clean-Up Kit, sufficient for 70 purifications
Sigma-Aldrich
SB-505124 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
MISSION® siRNA Product Offerings, Custom and Predesigned siRNA