Merck
  • Biomechanical properties of the thoracic aneurysmal wall: differences between bicuspid aortic valve and tricuspid aortic valve patients.
Biomechanical properties of the thoracic aneurysmal wall: differences between bicuspid aortic valve and tricuspid aortic valve patients.
The Annals of thoracic surgery (2014-06-03)
Caroline Forsell, Hanna M Björck, Per Eriksson, Anders Franco-Cereceda, T Christian Gasser
ABSTRACT
The prevalence for thoracic aortic aneurysms (TAAs) is significantly increased in patients with a bicuspid aortic valve (BAV) compared with patients who have a normal tricuspid aortic valve (TAV). TAA rupture is a life-threatening event, and biomechanics-based simulations of the aorta may help to disentangle the molecular mechanism behind its development and progression. The present study used polarized microscopy and macroscopic in vitro tensile testing to explore collagen organization and mechanical properties of TAA wall specimens from BAV and TAV patients. Circumferential sections of aneurysmal aortic tissue from BAV and TAV patients were obtained during elective operations. The distribution of collagen orientation was captured by a Bingham distribution, and finite element models were used to estimate constitutive model parameters from experimental load-displacement curves. Collagen orientation was almost identical in BAV and TAV patients, with a highest probability of alignment along the circumferential direction. The strength was almost two times higher in BAV samples (0.834 MPa) than in TAV samples (0.443 MPa; p<0.001). The collagen-related stiffness (Cf) was significantly increased in BAV compared with TAV patients (Cf=7.45 MPa vs 3.40 MPa; p=0.003), whereas the elastin-related stiffness was similar in both groups. A trend toward a decreased wall thickness was seen in BAV patients (p=0.058). The aneurysmal aortas of BAV patients show a higher macroscopic strength, mainly due to an increased collagen-related stiffness, compared with TAV patients. The increased wall stiffness in BAV patients may contribute to the higher prevalence for TAAs in this group.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance