Merck
  • The development and applications of sucralose, a new high-intensity sweetener.
The development and applications of sucralose, a new high-intensity sweetener.
Canadian journal of physiology and pharmacology (1994-04-01)
I Knight
ABSTRACT
Sucralose is a new sweetener discovered during a collaborative research program between Tate & Lyle and Queen Elizabeth College of the University of London. It is made by selective substitution of sucrose hydroxyl groups by chlorine, resulting in a highly intense (600x) sugarlike sweetness and exceptional stability at both high temperature and low pH. The research leading to the discovery of sweetness in differently halogenated sucrose is described, as well as the development of sucralose and the process of safety testing and government approval. Finally, sucralose properties and applications in Canada's food and beverage industries are discussed.
Product Description
Supelco
Sucralose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucralose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Supelco
Sucralose, analytical standard
Sucralose, European Pharmacopoeia (EP) Reference Standard