Merck
The disposition of saccharin in animals and man--a review.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1985-04-01)
A G Renwick
ABSTRACT
Recent studies on saccharin in animals and man have allowed a detailed understanding of its fate in the body. Saccharin is slowly absorbed from the gut but rapidly eliminated in the urine, largely by renal tubular secretion. Saccharin does not undergo detectable metabolism in either animals or man. Tissue specific accumulation in the urinary bladder, suggested by single and multiple dose studies in rats, was not found during chronic administration in the diet. The bladder tissue is part of the central, rapidly equilibrating, compartment. The sex- and generation-specificity of the tumorigenic effect is not due to unique accumulation in the urinary bladder of F1 males. Saturation of renal tubular secretion, which occurs in rats fed 5% saccharin or more in the diet, was not found in human volunteers given large oral doses (2 g).
Product Description
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
Saccharin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Saccharin, ≥99%
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)