- Selective reductive transformations using samarium diiodide-water.
Selective reductive transformations using samarium diiodide-water.
Chemical communications (Cambridge, England) (2011-09-01)
Michal Szostak, Malcolm Spain, Dixit Parmar, David J Procter
PMID21879049
ABSTRACT
Samarium diiodide (SmI(2)) is one of the most important reductive electron transfer reagents available in the laboratory. Key to the popularity of SmI(2) is the ability of additives and co-solvents to tune the properties of the reagent. Over the last decade water has emerged as a particularly valuable additive, opening up new chemical space and leading to the discovery of unprecedented selectivity and new reactions promoted by SmI(2). In this Feature Article we review recent progress in the application of SmI(2)-H(2)O systems, with an emphasis on mechanistic considerations and the development of new transformations.
Product Description
Samarium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Samarium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Samarium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Samarium, rod, 100mm, diameter 6.35mm, cast, 99%
Samarium, rod, 50mm, diameter 6.35mm, cast, 99%