Merck
  • Carcinogenicity of acetaldehyde in alcoholic beverages: risk assessment outside ethanol metabolism.
Carcinogenicity of acetaldehyde in alcoholic beverages: risk assessment outside ethanol metabolism.
Addiction (Abingdon, England) (2009-04-02)
Dirk W Lachenmeier, Fotis Kanteres, Jürgen Rehm
ABSTRACT
In addition to being produced in ethanol metabolism, acetaldehyde occurs naturally in alcoholic beverages. Limited epidemiological evidence points to acetaldehyde as an independent risk factor for cancer during alcohol consumption, in addition to the effects of ethanol. This study aims to estimate human exposure to acetaldehyde from alcoholic beverages and provide a quantitative risk assessment. The human dietary intake of acetaldehyde via alcoholic beverages was estimated based on World Health Organization (WHO) consumption data and literature on the acetaldehyde contents of different beverage groups (beer, wine, spirits and unrecorded alcohol). The risk assessment was conducted using the European Food Safety Authority's margin of exposure (MOE) approach with benchmark doses obtained from dose-response modelling of animal experiments. Life-time cancer risk was calculated using the T25 dose descriptor. The average exposure to acetaldehyde from alcoholic beverages was estimated at 0.112 mg/kg body weight/day. The MOE was calculated to be 498, and the life-time cancer risk at 7.6 in 10,000. Higher risk may exist for people exposed to high acetaldehyde contaminations, as we have found in certain unrecorded alcohol beverages in Guatemala and Russia, for which we have demonstrated possible exposure scenarios, with risks in the range of 1 in 1000. The life-time cancer risks for acetaldehyde from alcoholic beverages greatly exceed the usual limits for cancer risks from the environment set between 1 : 10,000 and 1 : 1,000,000. Alcohol consumption has thus been identified as a direct source of acetaldehyde exposure, which in conjunction with other sources (food flavourings, tobacco) results in a magnitude of risk requiring intervention. An initial public health measure could be to reduce the acetaldehyde content in alcoholic beverages as low as technologically possible, and to restrict its use as a food flavour additive.
Product Description
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, ≥99%, meets FCC analytical specification
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde Assay Kit, sufficient for 100 fluorometric tests
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF