- Novel bacterial molybdenum and tungsten enzymes: three-dimensional structure, spectroscopy, and reaction mechanism.
Novel bacterial molybdenum and tungsten enzymes: three-dimensional structure, spectroscopy, and reaction mechanism.
Biological chemistry (2005-10-13)
Matthias Boll, Bernhard Schink, Albrecht Messerschmidt, Peter M H Kroneck
PMID16218872
ABSTRACT
The molybdenum enzymes 4-hydroxybenzoyl-CoA reductase and pyrogallol-phloroglucinol transhydroxylase and the tungsten enzyme acetylene hydratase catalyze reductive dehydroxylation reactions, i.e., transhydroxylation between phenolic residues and the addition of water to a triple bond. Such activities are unusual for this class of enzymes, which carry either a mononuclear Mo or W center. Crystallization and subsequent structural analysis by high-resolution X-ray crystallography has helped to resolve the reaction centers of these enzymes to a degree that allows us to understand the interaction of the enzyme and the respective substrate(s) in detail, and to develop a concept for the respective reaction mechanism, at least in two cases.
Product Description
Tungsten, wire, straight, 300mm, diameter 0.30mm, 99.9+%
Tungsten, wire, straight, 300mm, diameter 0.75mm, 99.9+%
Tungsten, wire, straight, 25.4mm, diameter 0.5mm, 99.9+%
Tungsten, wire, straight, 300mm, diameter 0.125mm, 99.9+%
Tungsten, foil, 0.2m coil, thickness 0.02mm, coil width .1mm, as rolled, 99.95%
Tungsten, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.3mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.5mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.2mm, as rolled, 99.95%
Tungsten, foil, 200x200mm, thickness 0.15mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.08mm, coil width .9mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.10mm, coil width .9mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.035mm, coil width .5mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.2mm, as rolled, 99.95%
Tungsten, rod, 100mm, diameter 8.0mm, centerless ground, 99.95%
Tungsten, rod, 1000mm, diameter 3.0mm, centerless ground, 99.95%
Tungsten, wire reel, 0.5m, diameter 1.0mm, clean, 99.95%
Tungsten, rod, 200mm, diameter 6.35mm, centerless ground, 99.95%
Tungsten, microfoil, disks, 25mm, thinness 0.1μm, specific density 182μg/cm2, permanent mylar 3.5μm support, 99.95%
Tungsten, rod, 100mm, diameter 12.0mm, centerless ground, 99.95%
Tungsten, rod, 200mm, diameter 2.0mm, centerless ground, 99.95%
Tungsten, wire reel, 100m, diameter 0.05mm, gold plated, 99.95%
Tungsten, wire reel, 1000m, diameter 0.025mm, clean, 99.95%
Tungsten, rod, 25mm, diameter 20.0mm, centerless ground, 99.95%
Tungsten, wire reel, 100m, diameter 0.5mm, clean, 99.95%