Merck
Clinical pharmacokinetics of ketorolac tromethamine.
Clinical pharmacokinetics (1992-12-01)
D R Brocks, F Jamali
ABSTRACT
Ketorolac is a new chiral nonsteroidal anti-inflammatory drug (NSAID) which is marketed for analgesia as the racemate. The drug is administered as the water soluble tromethamine salt and is available in tablets or as an intramuscular injection. The absorption of ketorolac is rapid, Cmax being attained between 20 to 60 min. Its oral bioavailability is estimated to range from 80 to 100%. The drug is extensively bound (> 99%) to plasma proteins and has a volume of distribution (0.1 to 0.3 L/kg) comparable with those of other NSAIDs. Only small concentrations of ketorolac are detectable in umbilical vein blood after administration to women in labour. The elimination half-life is between 4 and 6h and is moderate in comparison with other NSAIDs. The area under the plasma concentration-time curve of ketorolac is proportional to the dose after intramuscular administration of therapeutic doses to young healthy volunteers. Ketorolac is extensively metabolised through glucuronidation and oxidation; little if any drug is eliminated unchanged. Most of the dose of ketorolac is recovered in the urine as conjugated drug. Although ketorolac is excreted into the breast milk, the amount of drug transferred comprises only a small fraction of the maternal exposure. Little stereoselectivity was present in the pharmacokinetics of ketorolac in a healthy volunteer receiving single intravenous or oral doses. The elderly exhibit reduced clearance of the drug. Renal insufficiency appears to cause an accumulation of ketorolac in plasma, although hepatic disease may not affect the pharmacokinetics.
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Supelco
Ketorolac Tromethamine, Pharmaceutical Secondary Standard; Certified Reference Material
Ketorolac trometamol for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Ketorolac tris salt, ≥99%, crystalline
Ketorolac trometamol, European Pharmacopoeia (EP) Reference Standard
SAFC
Tromethamine