Merck
Controversies in skin resurfacing: the role of erbium.
Journal of cutaneous laser therapy (2001-05-22)
D Fleming
ABSTRACT
Carbon dioxide laser resurfacing has been a valuable procedure for facial skin rejuvenation since the early 1990s, largely replacing medium and deep chemical peels and dermabrasion. The introduction of the erbium:YAG laser for resurfacing has caused confusion about its role. Because of its ability to resurface very superficially it has been limited by many laser surgeons to treating only superficial rhytides and sun damage. However, it is the equal of CO2 in improving deep rhytides but with quicker healing and fewer side-effects.
Product Description
Erbium, rod, 100mm, diameter 10.0mm, cast, 99%
Erbium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Erbium, rod, 100mm, diameter 6.35mm, cast, 99%
Erbium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Erbium, foil, 50x50mm, thickness 1.0mm, as rolled, 99%
Erbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Erbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Erbium, rod, 50mm, diameter 10.0mm, cast, 99%
Erbium, rod, 50mm, diameter 2.0mm, cast, 99%
Erbium, rod, 50mm, diameter 6.35mm, cast, 99%
Erbium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%