Merck
  • Beneficial effects of edaravone on the expression of serum matrix metalloproteinase-9 after cerebral hemorrhage.
Beneficial effects of edaravone on the expression of serum matrix metalloproteinase-9 after cerebral hemorrhage.
Neurosciences (Riyadh, Saudi Arabia) (2014-04-18)
Fahui Zhao, Zhigang Liu
ABSTRACT
To explore the effects of edaravone on the expression of matrix metalloproteinase-9 (MMP-9) posterior to cerebral hemorrhage, and to analyze the relationship between this expression and neurological deficit. A total of 160 basal ganglia hemorrhage patients enrolled in Dongfeng Hospital, Hubei University of Medicine, Shiyan, China between April 2009 and July 2011 were included and divided into a treatment group and a control group (n=80). All patients were treated with minimally invasive hematoma evacuation, and the treatment group was administered with edaravone simultaneously. Serum MMP-9 levels were measured by double-antibody sandwich enzyme-linked immunosorbent assay. The National Institutes of Health Stroke Scale (NIHSS) and Glasgow Coma Scale scores of the 2 groups were determined before and after treatment. The overall effective rates of the treatment (86.3%) and control (75.0%) groups differed significantly (p<0.05). The serum MMP-9 levels of the 2 groups that were similar (p>0.05) before treatment significantly decreased (p<0.05) after treatment, and those of the 2 groups also differed significantly (p<0.05). The NIHSS scores of the 2 groups that were similar (p>0.05) before treatment also significantly decreased (p<0.05) after treatment, and those of the 2 groups differed significantly (p<0.05). Pearson`s correlation analysis revealed that the level of serum MMP-9 was significantly correlated with the NIHSS score before treatment (R=0.491). Combined minimally invasive hematoma evacuation and edaravone effectively treated cerebral hemorrhage by directly lowering the level of serum MMP-9.
Sigma-Aldrich
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%
Supelco
Antipyrine, analytical standard
Sigma-Aldrich
3-Methyl-1-phenyl-2-pyrazoline-5-one
Phenazone, European Pharmacopoeia (EP) Reference Standard
Phenazone impurity A, European Pharmacopoeia (EP) Reference Standard