Merck
  • Synthesis and anti-staphylococcal activity of TiO2 nanoparticles and nanowires in ex vivo porcine skin model.
Synthesis and anti-staphylococcal activity of TiO2 nanoparticles and nanowires in ex vivo porcine skin model.
Journal of biomedical nanotechnology (2014-04-17)
Namrata Nataraj, G S Anjusree, Asha Anish Madhavan, P Priyanka, Deepthi Sankar, N Nisha, S V Lakshmi, R Jayakumar, Avinash Balakrishnan, Raja Biswas
ABSTRACT
Staphylococcus aureus is one of the major causes of skin and soft tissue infections. In this study we compared the antimicrobial activity of two different TiO2 nanoformulations against Staphylococcus aureus. We synthesized TiO2 nanoparticles of approximately 80 nm diameter and TiO2 nanowires of approximately 100 nm diameter. Both nanoformulations possess anti-microbial activity; were non-hemolytic and cytocompatible. However, the anti-staphylococcal activity of TiO2 nanowires was better than the nanoparticles. In broth culture, growth of S. aureus was only partially inhibited by 2% and 4 wt% TiO2 nanoparticles and completely inhibited by TiO2 nanowires till 24 h. TiO2 nanowires treated S. aureus cells exhibits diminished membrane potential than nanoparticle treated cells. The anti-microbial properties of both TiO2 nanoformulations were validated using ex vivo porcine skin model which supplements the in vitro assays. Anti-bacterial activity of the TiO2 nanowires were also validated against multi drug resistant pathogenic strains of S. aureus, showing the clinical potency of the TiO2 nanowires compared to its nanoparticles.
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Titanium, wire reel, 5m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 1.0mm, as drawn, 99.6+%
Titanium, tube, 1000mm, outside diameter 4.25mm, inside diameter 3.75mm, wall thickness 0.25mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, tube, 1220mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 4.25mm, inside diameter 3.75mm, wall thickness 0.25mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 8.0mm, as drawn, 99.99+%
Titanium, rod, 200mm, diameter 9.5mm, annealed, 99.99+%
Titanium, rod, 100mm, diameter 9.5mm, annealed, 99.99+%
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 2mm, as drawn, 99.99+%
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%