Merck
IL-6 alters osteocyte signaling toward osteoblasts but not osteoclasts.
Journal of dental research (2014-02-05)
A D Bakker, R N Kulkarni, J Klein-Nulend, W F Lems
ABSTRACT
Mechanosensitive osteocytes regulate bone mass in adults. Interleukin 6 (IL-6), such as present during orthodontic tooth movement, also strongly affects bone mass, but little is known about the effect of IL-6 on osteocyte function. Therefore we aimed to determine in vitro whether IL-6 affects osteocyte mechanosensitivity, and osteocyte regulation of osteoclastogenesis and osteoblast differentiation. MLO-Y4 osteocytes were incubated with/without IL-6 (1 or 10 pg/mL) for 24 hr. Subsequently, osteocytes were subjected to mechanical loading by pulsating fluid flow (PFF) for 1 hr. Mouse osteoclast precursors were cultured for 7 days on top of IL-6-treated osteocytes. Conditioned medium from osteocytes treated with/without IL-6 was added to MC3T3-E1 pre-osteoblasts for 14 days. Exogenous IL-6 (10 pg/mL) did not alter the osteocyte response to PFF. PFF significantly enhanced IL-6 production by osteocytes. IL-6 enhanced Rankl expression but reduced caspase 3/7 activity by osteocytes, and therefore did not affect osteocyte-stimulated osteoclastogenesis. Conditioned medium from IL-6-treated osteocytes reduced alkaline phosphatase (ALP) activity and Runx2 expression in osteoblasts, but increased expression of the proliferation marker Ki67 and osteocalcin. Our results suggest that IL-6 is produced by shear-loaded osteocytes and that IL-6 may affect bone mass by modulating osteocyte communication toward osteoblasts.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host