Merck
  • In vitro Characterization of a small molecule inhibitor of the alanine serine cysteine transporter -1 (SLC7A10).
In vitro Characterization of a small molecule inhibitor of the alanine serine cysteine transporter -1 (SLC7A10).
Journal of neurochemistry (2013-11-26)
Jeffrey M Brown, Lisa Hunihan, Margaret M Prack, David G Harden, Joanne Bronson, Carolyn D Dzierba, Robert G Gentles, Adam Hendricson, Rudy Krause, John E Macor, Ryan S Westphal
ABSTRACT
NMDA receptor hypofunction is hypothesized to contribute to cognitive deficits associated with schizophrenia. Since direct activation of NMDA receptors is associated with serious adverse effects, modulation of the NMDA co-agonists, glycine or D-serine, represents a viable alternative therapeutic approach. Indeed, clinical trials with glycine and D-serine have shown positive results, although concerns over toxicity related to the high-doses required for efficacy remain. Synaptic concentrations of D-serine and glycine are regulated by the amino acid transporter alanine serine cysteine transporter-1 (asc-1). Inhibition of asc-1 would increase synaptic D-serine and possibly glycine, eliminating the need for high-dose systemic D-serine or glycine treatment. In this manuscript, we characterize Compound 1 (BMS-466442), the first known small molecule inhibitor of asc-1. Compound 1 selectively inhibited asc-1 mediated D-serine uptake with nanomolar potency in multiple cellular systems. Moreover, Compound 1 inhibited asc-1 but was not a competitive substrate for this transporter. Compound 1 is the first reported selective inhibitor of the asc-1 transporter and may provide a new path for the development of asc-1 inhibitors for the treatment of schizophrenia.
Sigma-Aldrich
BMS-466442, ≥98% (HPLC)
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
SAFC
L-Histidine
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Histidine, European Pharmacopoeia (EP) Reference Standard