Merck
  • Engineered zinc-finger transcription factors activate OCT4 (POU5F1), SOX2, KLF4, c-MYC (MYC) and miR302/367.
Engineered zinc-finger transcription factors activate OCT4 (POU5F1), SOX2, KLF4, c-MYC (MYC) and miR302/367.
Nucleic acids research (2014-05-06)
Qingzhou Ji, Ashley L Fischer, Clyde R Brown, Erik R Eastlund, Tamar Dvash, Bonan Zhong, Mark A Gerber, Ian Lyons, Scott W Knight, Carol A Kreader
ABSTRACT
Artificial transcription factors are powerful tools for regulating gene expression. Here we report results with engineered zinc-finger transcription factors (ZF-TFs) targeting four protein-coding genes, OCT4, SOX2, KLF4 and c-MYC, and one noncoding ribonucleic acid (RNA) gene, the microRNA (miRNA) miR302/367 cluster. We designed over 300 ZF-TFs whose targets lie within 1 kb of the transcriptional start sites (TSSs), screened them for increased messenger RNA or miRNA levels in transfected cells, and identified potent ZF-TF activators for each gene. Furthermore, we demonstrate that selected ZF-TFs function with alternative activation domains and in multiple cell lines. For OCT4, we expanded the target range to -2.5 kb and +500 bp relative to the TSS and identified additional active ZF-TFs, including three highly active ZF-TFs targeting distal enhancer, proximal enhancer and downstream from the proximal promoter. Chromatin immunoprecipitation (FLAG-ChIP) results indicate that several inactive ZF-TFs targeting within the same regulatory region bind as well as the most active ZF-TFs, suggesting that efficient binding within one of these regulatory regions may be necessary but not sufficient for activation. These results further our understanding of ZF-TF design principles and corroborate the use of ZF-TFs targeting enhancers and downstream from the TSS for transcriptional activation.
Sigma-Aldrich
KiCqStart® SYBR® Green qPCR ReadyMix, iQ, with fluorescein for Bio-Rad systems
Sigma-Aldrich
KiCqStart® SYBR® Green qPCR ReadyMix, For Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems
Sigma-Aldrich
KiCqStart® SYBR® Green qPCR ReadyMix, Low ROX, for ABI and Stratagene instruments
Sigma-Aldrich
SYBR® Green Quantitative RT-qPCR Kit, One step SYBR® Green RT-qPCR with MMLV & hot-start Taq DNA Polymerase
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Protein A Mag Sepharose Xtra, Cytiva 28-9670-56, pack of 2 × 1 mL