Merck
  • Use of a glyphosate-based herbicide-induced nephrotoxicity model to investigate a panel of kidney injury biomarkers.
Use of a glyphosate-based herbicide-induced nephrotoxicity model to investigate a panel of kidney injury biomarkers.
Toxicology letters (2013-12-24)
Klintean Wunnapuk, Glenda Gobe, Zoltan Endre, Philip Peake, Jeffrey E Grice, Michael S Roberts, Nicholas A Buckley, Xin Liu
ABSTRACT
Accidental or intentional ingestion of glyphosate surfactant-based herbicides, like Roundup(®), leads to nephrotoxicity as well as death. In this study, a panel of kidney injury biomarkers was evaluated in terms of suitability to detect acute kidney injury and dysfunction. The Roundup(®) intoxication model involved oral administration of glyphosate to rats at dose levels of 250, 500, 1200 and 2500 mg/kg. Urinary and plasma biomarker patterns were investigated at 8, 24 and 48 h after dosing. Biomarkers were quantified by absolute concentration; by normalising to urine creatinine; and by calculating the excretion rate. The diagnostic performances of each method in predicting of acute kidney injury were compared. By Receiver Operating Characteristic (ROC) analysis of the selected biomarkers, only urinary kidney injury molecule-1 (KIM-1) best predicted histological changes at 8h (best cut-off point>0.00029 μg/ml). Plasma creatinine performed better than other biomarkers at 24 h (best cut-off point>0.21 mg/dl). Urinary KIM-1 was the best early biomarker of kidney injury in this glyphosate-induced nephrotoxicity model.
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
MILLIPLEX® Rat Kidney Toxicity Magnetic Bead Panel 1 - Toxicity Multiplex Assay, The analytes available for this multiplex kit are: Clusterin, GSTα, IP-10, KIM-1, OPN - Osteopontin, TIMP-1, VEGF.
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Supelco
Glyphosate, PESTANAL®, analytical standard
Millipore
MILLIPLEX® Rat Kidney Toxicity Magnetic Bead Panel 2 - Toxicity Multiplex Assay, The analytes available for this multiplex kit are: Albumin, AGP, ß2M, Cystatin C, EGF, Lipocalin-2/NGAL.
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)