Merck
  • Branched-chain amino acid requirements for enterally fed term neonates in the first month of life.
Branched-chain amino acid requirements for enterally fed term neonates in the first month of life.
The American journal of clinical nutrition (2013-11-29)
Femke de Groof, Lisha Huang, Ineke van Vliet, Gardi J Voortman, Henk Schierbeek, Lodi C W Roksnoer, Andras Vermes, Chao Chen, Ying Huang, Johannes B van Goudoever
ABSTRACT
Knowledge of essential amino acid requirements in infants is important because excessive intake of protein can lead to increased long-term morbidity such as obesity. A deficient intake may lead to suboptimal growth and impaired neurodevelopment. The current recommended branched-chain amino acid requirements in infants aged 0-1 mo are based on the amino acid content of human milk. We quantified the requirements for isoleucine, leucine, and valine for term neonates by using the indicator amino acid oxidation method with [1-(13)C]phenylalanine as the indicator. Fully enterally fed term infants received randomly graded amounts of isoleucine (5-216 mg · kg(-1) · d(-1)), leucine (5-370 mg · kg(-1) · d(-1)), or valine (5-236 mg · kg(-1) · d(-1)) as part of an elemental formula. Data are expressed as means ± SDs. Eighty-three Asian, term neonates (mean ± SD birth weight: 3.3 ± 0.4 kg; gestational age: 39.4 ± 1.3 wk) were studied at a postnatal age of 13 ± 5 d. Mean requirements for isoleucine, leucine, and valine (measured in boys only) were 105 mg · kg(-1) · d(-1) (r(2) = 0.61, P < 0.001), 140 mg · kg(-1) · d(-1) (r(2) = 0.26, P < 0.01), and 110 mg · kg(-1) · d(-1) (r(2) = 0.35, P = 0.001), respectively. Current human milk-based recommendations for isoleucine and valine in term infants aged 0-1 mo are correct. However, the current recommendation for leucine (166 mg · kg(-1) · d(-1)) is higher than the mean requirement of 140 mg · kg(-1) · d(-1) that we determined in this study. This trial was registered at www.trialregister.nl as NTR1610.
Product Description
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
SAFC
L-Valine
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
SAFC
L-Isoleucine