Merck
  • Unsynchronised subunit motion in single trimeric sodium-coupled aspartate transporters.
Unsynchronised subunit motion in single trimeric sodium-coupled aspartate transporters.
Nature (2013-10-05)
Guus B Erkens, Inga Hänelt, Joris M H Goudsmits, Dirk Jan Slotboom, Antoine M van Oijen
ABSTRACT
Excitatory amino acid transporters (EAATs) are secondary transport proteins that mediate the uptake of glutamate and other amino acids. EAATs fulfil an important role in neuronal signal transmission by clearing the excitatory neurotransmitters from the synaptic cleft after depolarization of the postsynaptic neuron. An intensively studied model system for understanding the transport mechanism of EAATs is the archaeal aspartate transporter GltPh. Each subunit in the homotrimeric GltPh supports the coupled translocation of one aspartate molecule and three Na(+) ions as well as an uncoupled flux of Cl(-) ions. Recent crystal structures of GltPh revealed three possible conformations for the subunits, but it is unclear whether the motions of individual subunits are coordinated to support transport. Here, we report the direct observation of conformational dynamics in individual GltPh trimers embedded in the membrane by applying single-molecule fluorescence resonance energy transfer (FRET). By analysing the transporters in a lipid bilayer instead of commonly used detergent micelles, we achieve conditions that approximate the physiologically relevant ones. From the kinetics of FRET level transitions we conclude that the three GltPh subunits undergo conformational changes stochastically and independently of each other.
Product Description
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
SAFC
L-Aspartic acid
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Pricing and availability is not currently available.