Merck
  • Effect of extrusion cooking on the physicochemical properties, resistant starch, phenolic content and antioxidant capacities of green banana flour.
Effect of extrusion cooking on the physicochemical properties, resistant starch, phenolic content and antioxidant capacities of green banana flour.
Food chemistry (2013-09-24)
Chonthira Sarawong, Regine Schoenlechner, Ken Sekiguchi, Emmerich Berghofer, Perry K W Ng
ABSTRACT
Green banana flour was extruded through a co-rotating twin-screw extruder with constant barrel temperature. The objectives of this study were to determine the effect of extrusion cooking variables (feed moisture, FM, 20% and 50%; screw speed, SS, 200 and 400rpm) and storing of the extruded flours at 4°C for 24h on the physicochemical properties, resistant starch (RS), pasting properties and antioxidant capacities. Extrusion cooking at higher FM and lower SS increased the amylose content, which was expressed in highest RS content. Water adsorption index (WAI) and pasting properties were increased, while water solubility index (WSI), total phenolic content (TPC) and antioxidant activities (FRAP, ABTS(+), DPPH) in free and bound phenolics were decreased compared to the other extruded samples. Storing the extruded flours at 4°C for 24h prior to oven drying was the main factor leading to a further increase in the content of amylose, RS, TPC and WSI values, as well as pasting properties - in particular peak viscosity. Compared to native banana flour, extrusion cooking caused significant changes in all studied properties of the extruded flours, except for soluble DF and antioxidant capacity (ABTS(+) and DPPH) of bound phenolics.
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, Powder
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch from corn