Merck
  • Peroxidase-type reactions suggest a heterolytic/nucleophilic O-O joining mechanism in the heme-dependent chlorite dismutase.
Peroxidase-type reactions suggest a heterolytic/nucleophilic O-O joining mechanism in the heme-dependent chlorite dismutase.
Biochemistry (2013-09-05)
Jeffrey A Mayfield, Béatrice Blanc, Kenton R Rodgers, Gudrun S Lukat-Rodgers, Jennifer L DuBois
ABSTRACT
Heme-containing chlorite dismutases (Clds) catalyze a highly unusual O-O bond-forming reaction. The O-O cleaving reactions of hydrogen peroxide and peracetic acid (PAA) with the Cld from Dechloromonas aromatica (DaCld) were studied to better understand the Cl-O cleavage of the natural substrate and subsequent O-O bond formation. While reactions with H2O2 result in slow destruction of the heme, at acidic pH heterolytic cleavage of the O-O bond of PAA cleanly yields the ferryl porphyrin cation radical (compound I). At alkaline pH, the reaction proceeds more rapidly, and the first observed intermediate is a ferryl heme. Freeze-quench EPR confirmed that the latter has an uncoupled protein-based radical, indicating that compound I is the first intermediate formed at all pH values and that radical migration is faster at alkaline pH. These results suggest by analogy that two-electron Cl-O bond cleavage to yield a ferryl-porphyrin cation radical is the most likely initial step in O-O bond formation from chlorite.
Sigma-Aldrich
Peracetic acid solution, 8.7% in acetic acid
Sigma-Aldrich
Peracetic acid solution, 32 wt. % in dilute acetic acid