Merck
  • Fatty acid-induced mitochondrial uncoupling elicits inflammasome-independent IL-1α and sterile vascular inflammation in atherosclerosis.
Fatty acid-induced mitochondrial uncoupling elicits inflammasome-independent IL-1α and sterile vascular inflammation in atherosclerosis.
Nature immunology (2013-09-03)
Stefan Freigang, Franziska Ampenberger, Adrienne Weiss, Thirumala-Devi Kanneganti, Yoichiro Iwakura, Martin Hersberger, Manfred Kopf
ABSTRACT
Chronic inflammation is a fundamental aspect of metabolic disorders such as obesity, diabetes and cardiovascular disease. Cholesterol crystals are metabolic signals that trigger sterile inflammation in atherosclerosis, presumably by activating inflammasomes for IL-1β production. We found here that atherogenesis was mediated by IL-1α and we identified fatty acids as potent inducers of IL-1α-driven vascular inflammation. Fatty acids selectively stimulated the release of IL-1α but not of IL-1β by uncoupling mitochondrial respiration. Fatty acid-induced mitochondrial uncoupling abrogated IL-1β secretion, which deviated the cholesterol crystal-elicited response toward selective production of IL-1α. Our findings delineate a previously unknown pathway for vascular immunopathology that links the cellular response to metabolic stress with innate inflammation, and suggest that IL-1α, not IL-1β, should be targeted in patients with cardiovascular disease.
Product Description
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Supelco
Oleic acid, analytical standard